3-Aminophenylboronic acid 98 (contains varying amounts of Anhydride)
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
Dihydroxy(3-aminophenyl)borane, (3-Aminophenyl)boronic acid, 3-Aminobenzeneboronic acid, (3-Aminophenyl)metaboric acid
A135767-100G
A135767-1G
ALA135767-100GEA
949.91
USD
ALA135767-100G
ALA135767-1G
3-Aminophenylboronic acid 98 (contains varying amounts of Anhydride)
3-Aminophenylboronic acid
A boronic acid with potential use for biochemical research
Formula:
C₆H₈BNO₂ MW: 136.95 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00007755 CAS Number: 30418-59-8 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...