Allyl cinnamate ≥99%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: Allyl Cinnamate, Cinnamic Acid Allyl Ester, Cinnamic acid Allyl Ester

A135568-100G A135568-25G
ALA135568-100GEA 72.11 USD
ALA135568-100G ALA135568-25G
Allyl cinnamate ≥99%
Allyl cinnamate
Formula: C₁₂H₁₂O₂
MW: 188.23 g/mol
Flash Pt: 156°C
MDL Number: MFCD00026105
CAS Number: 1866-31-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a7a89f1c1eb45635e787ab8760d03bdf174890fe&vid=a7a89f1c1eb45635e787ab8760d03bdf174890fe&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR