2'-Aminoacetanilide ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A135471-5G A135471-100G A135471-1G A135471-250MG A135471-25G
ALA135471-5GEA 59.76 USD
ALA135471-5G ALA135471-100G ALA135471-1G ALA135471250MG ALA135471-25G
2'-Aminoacetanilide ≥98%
2'-Aminoacetanilide

Product Introduction:2′-Aminoacetanilide (2-aminoacetanilide, o-aminoacetanilide) can be prepared by the catalytic hydrogenation of 2-nitroacetanilide using 10%Pd/C (palladium/carbon).application:2′-Aminoacetanilide may be used in the preparation of:2-Methylbenzimidazole.N-(2-(1,3-Dimethyl-2,4-dioxo-5-phenyl-3,4-dihydro-1H-pyrrolo[3,4-d]pyrimidin-6(2H)-yl)phenyl)-5-methylfuran-2-carboxamide, a potent cystic fibrosis transmembrane conductance regulator (CFTR) inhibitor.Azobenzothiazole dyes, N-[2-(6-nitrobenzothiazol-2-ylazo)phenyl]acetamide and N-[2-(benzothiazol-2-ylazo)phenyl]acetamide.

Formula: C₈H₁₀N₂O
MW: 150.18
Melting Pt: 133-137°C
MDL Number: MFCD00210388
CAS Number: 34801-09-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e5610a755fed13c1f12a2bda38d1299e92a8684f&vid=e5610a755fed13c1f12a2bda38d1299e92a8684f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR