2-Amino-3,5-dichloro-6-methylpyridine ≥97% (by titration)

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 6-Amino-3,5-dichloro-2-methylpyridine

A135368-100G A135368-1G A135368-200MG A135368-25G A135368-5G
ALA135368-100GEA 570.86 USD
ALA135368-100G ALA135368-1G ALA135368200MG ALA135368-25G ALA135368-5G
2-Amino-3,5-dichloro-6-methylpyridine ≥97% (by titration)
2-Amino-3,5-dichloro-6-methylpyridine
Formula: C₆H₆Cl₂N₂
Melting Pt: 134 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00129029
CAS Number: 22137-52-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2c1f9e518cc4567f5bd76ce0bca53d56130742ca&vid=2c1f9e518cc4567f5bd76ce0bca53d56130742ca&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR