Ethyl-4-amino-2-(methylthio)pyrimidine-5-carboxylate ≥95%
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
Ethyl 4-amino-2-(methylthio)pyrimidin-5-carboxylate, Ethyl 4-amino-2-(methylthio)pyrimidine-5-carboxylate
A135301-100G
A135301-1G
A135301-250MG
A135301-5G
ALA135301-100GEA
713.36
USD
ALA135301-100G
ALA135301-1G
ALA135301250MG
ALA135301-5G
Ethyl-4-amino-2-(methylthio)pyrimidine-5-carboxylate ≥95%
Ethyl 4-amino-2-(methylthio)pyrimidine-5-carboxylate
Formula:
C₈H₁₁N₃O₂S |
MDL Number:
MFCD00090781 CAS Number: 776-53-4 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...