4-Amino-1,3,5-trimethyl-1H-pyrazole ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 1,3,5-Trimethyl-1H-pyrazol-4-amine

A134908-1G A134908-25G A134908-5G
ALA134908-1GEA 74.01 USD
ALA134908-1G ALA134908-25G ALA134908-5G
4-Amino-1,3,5-trimethyl-1H-pyrazole ≥97%
4-Amino-1,3,5-trimethyl-1H-pyrazole
Formula: C₆H₁₁N₃
MW: 125.17 g/mol
Melting Pt: 99-105°C
MDL Number: MFCD00052883
CAS Number: 28466-21-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7149a127b884cd082c55e74c2b3fef2069d9bc27&vid=7149a127b884cd082c55e74c2b3fef2069d9bc27&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR