Anisotropine Methylbromide ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: Anisotropine Methobromide, Vapin, 8-Methyltropinium Bromide 2-Propylpentanoate, 8-Methyltropinium Bromide, 2-Propylvalerate, endo-8,8-Dimethyl-3-[(1-oxo-2-propylpentyl)oxy]- 8-azoniabicyclo[3.2.1]octane Bromide, (3-endo)-8,8-Dimethyl-3-[(1-oxo-2-propylpentyl)oxy]-8-azoniabicyclo[3.2.1]octane Bromide, 8-Methyl-3-(2-propylpentanoyloxy)tropinium Bromide, Lytispasm, Valpin, Octatropine Methylbromide, Valpin 50, 3α-Hydroxy-8-methyl-1αH,5αH-tropanium Bromide 2-Propylvalerate

A134744-5MG A134744-100MG A134744-10MG A134744-25MG A134744-50MG
ALA134744-5MGEA 66.41 USD
ALA134744-5MG ALA134744100MG ALA134744-10MG ALA134744-25MG ALA134744-50MG
Anisotropine Methylbromide ≥98%
Anisotropine Methylbromide
Formula: C₁₇H₃₂BrNO₂
Melting Pt: >285°C
Storage Temperature: Freezer
MDL Number: MFCD00083234
CAS Number: 80-50-2
EINECS: 201-285-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9b9ef9a63416402c05d2e1ce58e9c3726719b1d3&vid=9b9ef9a63416402c05d2e1ce58e9c3726719b1d3&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR