Isonicotinamidine hydrochloride ≥98%
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
Pyridine-4-carboxamidine hydrochloride, Isonicotinimidamide hydrochloride, 4-Carbamimidoylpyridine hydrochloride, 4-Amidinopyridine hydrochloride, Pyridine-4-carboximidamide hydrochloride, 4-Amidinopyridinium chloride
A134096-100G
A134096-1G
A134096-250MG
A134096-25G
A134096-5G
ALA134096-100GEA
1195.96
USD
ALA134096-100G
ALA134096-1G
ALA134096250MG
ALA134096-25G
ALA134096-5G
Isonicotinamidine hydrochloride ≥98%
Isonicotinamidine hydrochloride
Formula:
C₆H₇N₃·ClH Melting Pt: 245°C |
MDL Number:
MFCD00040385 CAS Number: 6345-27-3 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...