2-Amino-3-fluoro-5-(trifluoromethyl)pyridine ≥97%

Supplier: ALADDIN SCIENTIFIC

Synonyms: 3-Fluoro-5-(trifluoromethyl)pyridin-2-amine

A134043-1G A134043-250MG
ALA134043-1GEA 118.66 USD
ALA134043-1G ALA134043250MG
2-Amino-3-fluoro-5-(trifluoromethyl)pyridine ≥97%
2-Amino-3-fluoro-5-(trifluoromethyl)pyridine
Formula: C₆H₄F₄N₂
MW: 180.10 g/mol
MDL Number: MFCD10699117
CAS Number: 852062-17-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f543d2b5fd614fcbfe58def3322a0c6947d4783f&vid=f543d2b5fd614fcbfe58def3322a0c6947d4783f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR