Aminoacetamidine dihydrobromide ≥95%
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
A133988-1G
A133988-250MG
ALA133988-1GEA
115.81
USD
ALA133988-1G
ALA133988250MG
Aminoacetamidine dihydrobromide ≥95%
Aminoacetamidine dihydrobromide
2-Aminoacetamidine dihydrobromide may be used in the synthesis of 2-amino-5-(4-(2-(N-benzyloxycarbonylamino)ethoxy)phenyl)pyradine.
Formula:
C₂H₇N₃ Melting Pt: 227-232°C Storage Temperature: Ambient |
MDL Number:
MFCD00671486 CAS Number: 69816-37-1 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...