1-Ethylpropylamine ≥98% (by GC)

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: 3-Pentylamine, 3-Aminopentane, Pentan-3-amine

A133949-100ML A133949-25ML A133949-5ML
ALA133949100MLEA 158.56 USD
ALA133949100ML ALA133949-25ML ALA133949-5ML
1-Ethylpropylamine ≥98% (by GC)
1-Ethylpropylamine

It has been used in the synthesis of (1-ethyl-propyl)-[3-(4-methoxy-2-methylphenyl)-2-methyl-6,7-dihydro-5H-1,4,8a-triaza-s-indacen-8-yl]-amine, in the synthesis of monoclinic VPO4·H2O, as internal standard in the screening of aziridine and 2-chloroethylamine in active pharmaceutical ingredients by gas chromatography-mass spectrometry.

Formula: C₅H₁₃N
MW: 87.16 g/mol
Boiling Pt: 90
Flash Pt: 2°C
Storage Temperature: Ambient
MDL Number: MFCD00008096
CAS Number: 616-24-0
UN: 2733
ADR: 8(3) II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ce4252e67008d7df27523b7af712ad0a3de92be8&vid=ce4252e67008d7df27523b7af712ad0a3de92be8&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR