4-Amino-2,6-dimethoxypyrimidine ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 6-Amino-2,4-dimethoxypyrimidine, 4-Amino-2,6-dimethoxypyrimidine

A133799-100G A133799-25G A133799-5G
ALA133799-100GEA 205.11 USD
ALA133799-100G ALA133799-25G ALA133799-5G
4-Amino-2,6-dimethoxypyrimidine ≥98%
4-Amino-2,6-dimethoxypyrimidine

It is methoxy substituted 4-aminopyrimidine.Molecules of 4-amino-2,6-dimethoxypyrimidine are linked by an N-H.O hydrogen bond and an N-H.N hydrogen bond, forming sheets containing centrosymmetric rings.Photocatalytic degradation of 4-amino-2,6-dimethoxypyrimidine on TiO2 has been reported. Mass spectra of 4-amino-2,6-dimethoxypyrimidine has been studied.

Formula: C₆H₉N₃O₂
Melting Pt: 151 °C
MDL Number: MFCD00082802
CAS Number: 3289-50-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cc20e826664087af97f1eeb200cc558bf4bb436f&vid=cc20e826664087af97f1eeb200cc558bf4bb436f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR