(2-Aminophenyl)acetonitrile ≥97%

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: 2-(2-Aminophenyl)acetonitrile, 2-Aminobenzyl cyanide, (2-Aminophenyl)acetonitrile

A133628-1G A133628-250MG A133628-5G
ALA133628-1GEA 188.01 USD
ALA133628-1G ALA133628250MG ALA133628-5G
(2-Aminophenyl)acetonitrile ≥97%
(2-Aminophenyl)acetonitrile

It may be used in the preparation of oxindole.

Formula: C₈H₈N₂
MW: 132.17 g/mol
Melting Pt: 69-73°C
MDL Number: MFCD00091082
CAS Number: 2973-50-4
EINECS: 221-015-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e2df0a94e7facf3789d830d4090c441f21f1f5e1&vid=e2df0a94e7facf3789d830d4090c441f21f1f5e1&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR