4-Amino-6-chloro-1,3-benzenedisulphonyl Dichloride ≥98%

Supplier: ALADDIN SCIENTIFIC

Synonyms: 4-Amino-6-Chloro-Benzene-1,3-Disulfonyl Chloride, 4-Amino-6-Chlorobenzene-1,3-Di(Sulphonyl Chloride)

A133549-1G A133549-250MG
ALA133549-1GEA 318.16 USD
ALA133549-1G ALA133549250MG
4-Amino-6-chloro-1,3-benzenedisulphonyl Dichloride ≥98%
4-Amino-6-chloro-1,3-benzenedisulphonyl Dichloride
Formula: C₆H₄Cl₃NO₄S₂
CAS Number: 671-89-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8f62d3a363b895c303cb08cd8551c07cc0da089c&vid=8f62d3a363b895c303cb08cd8551c07cc0da089c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR