2-Amino-4-chloro-5-methylbenzenesulfonic acid sodium salt ≥98% (by HPLC)

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 5-Chloro-4-methylaniline-2-sulphonic acid Sodium Salt, Sodium 2-amino-4-chloro-5-methylbenzenesulfonate, Sodium 2-Amino-4-chloro-5-methylbenzenesulfonate, 5-Chloro-4-methylaniline-2-sulfonic acid Sodium Salt, Aminochlorotoluenesulfonicacidsodiumsalt, Sodium 2-amino-4-chloro-5-methylbenzenesulphonate, Aminochlorotoluenesulphonicacidsodiumsalt, 5-Chloro-p-toluidine-2-sulphonic acid Sodium Salt, Sodium 2-Amino-4-chloro-5-methylbenzenesulphonate, 5-Chloro-p-toluidine-2-sulfonic acid Sodium Salt

A133545-100G A133545-25G
ALA133545-100GEA 49.31 USD
ALA133545-100G ALA133545-25G
2-Amino-4-chloro-5-methylbenzenesulfonic acid sodium salt ≥98% (by HPLC)
2-Amino-4-chloro-5-methylbenzenesulphonic acid sodium salt
Formula: C₇H₇ClNNaO₃S
MDL Number: MFCD00070551
CAS Number: 6627-59-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e4cfd2b173bf3e360e3913b40df6ce82acded5b5&vid=e4cfd2b173bf3e360e3913b40df6ce82acded5b5&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR