4-Amylcyclohexanol (cis- and trans- mixture) ≥98% (by GC)

Supplier: ALADDIN SCIENTIFIC

Synonyms: 4-Pentylcyclohexanol

A133487-100G A133487-25G A133487-5G
ALA133487-100GEA 975.56 USD
ALA133487-100G ALA133487-25G ALA133487-5G
4-Amylcyclohexanol (cis- and trans- mixture) ≥98% (by GC)
4-Amylcyclohexanol (cis- and trans- mixture)
Formula: C₁₁H₂₂O
MW: 170.30 g/mol
MDL Number: MFCD01017412
CAS Number: 54410-90-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1c23a9a97c6f13389296844c16ad0b8680caa055&vid=1c23a9a97c6f13389296844c16ad0b8680caa055&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR