2-Amino-5-chlorobenzamide 98

Supplier: ALADDIN SCIENTIFIC
Warning

A133469-100G A133469-1G A133469-25G A133469-5G
ALA133469-100GEA 1155.11 USD
ALA133469-100G ALA133469-1G ALA133469-25G ALA133469-5G
2-Amino-5-chlorobenzamide 98
2-Amino-5-chlorobenzamide

Application:2-Amino-5-chlorobenzamide react with 2-Azido-benzoyl chloride to get 2-Azido-5'-chloro-2'-carboxamidobenzanilide.

Formula: C₇H₇ClN₂O
MW: 170.60 g/mol
Melting Pt: 171 °C
MDL Number: MFCD00017126
CAS Number: 5202-85-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=724eed58f738ad30756fd82458af1287f4da902b&vid=724eed58f738ad30756fd82458af1287f4da902b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR