2-Amino-4-chlorobenzonitrile ≥98%

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: 5-Chloro-2-cyanoaniline

A133325-100G A133325-1G A133325-5G
ALA133325-100GEA 652.56 USD
ALA133325-100G ALA133325-1G ALA133325-5G
2-Amino-4-chlorobenzonitrile ≥98%
2-Amino-4-chlorobenzonitrile

2-Amino-4-chlorobenzonitrile (2A4CBN) is a benzonitrile derivative. A study of the molecular structure, vibrational spectra and NBO analysis of 2A4CBN has been undertaken.Transformation of 2A4CBN into quinazoline-2,4(1H,3H)-diones at atmospheric pressure of CO2 in the presence of monomeric tungstate, TBA2[WO4] (TBA = tetra-n-butylammonium) has been reported.2-Amino-4-chlorobenzonitrile may be used in the preparation of the following:6-chloro-4-(1-diethylamino-4-pentylamino)-2-(p-methoxyphenyl)-quinazoline dihydrochloride;7-chloro-4-(1-diethylamino-4-pentylamino)-2-(p-methoxyphenyl)-quinazoline dihydrochloride;4,7-dichloro-2-(2-methylprop-1-enyl)-6-nitroquinazoline.6-chlorotacrine;N-(2-chloro-5-cyanophenyl)-4,4,4-trifluorobutanamide.

Formula: C₇H₅ClN₂
MW: 152.58
Melting Pt: 163 °C
MDL Number: MFCD00035927
CAS Number: 38487-86-4
EINECS: 253-967-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=23c258363cc6bf27428c5ef6c7d556c43c66f26e&vid=23c258363cc6bf27428c5ef6c7d556c43c66f26e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR