3-Amino-4-bromo-1H-pyrazole ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 4-Bromo-1H-pyrazol-5-amine, 4-Bromo-1H-pyrazol-3-amine, 3-Amino-4-bromopyrazole

A133246-100G A133246-10G A133246-1G A133246-25G A133246-5G
ALA133246-100GEA 1184.56 USD
ALA133246-100G ALA133246-10G ALA133246-1G ALA133246-25G ALA133246-5G
3-Amino-4-bromo-1H-pyrazole ≥97%
3-Amino-4-bromo-1H-pyrazole
Formula: C₃H₄BrN₃
Melting Pt: 138 °C
MDL Number: MFCD00082728
CAS Number: 16461-94-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1fcfdf5f97e6693dc12a2e54eab740a4a5396853&vid=1fcfdf5f97e6693dc12a2e54eab740a4a5396853&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR