Toluene-α-sulfonyl chloride ≥97%

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: Phenylmethylsulfonyl chloride, Phenylmethanesulfonyl chloride, Benzylsulphonyl chloride, α-Toluenesulfonyl chloride, α-Toluenesulphonyl chloride, Phenylmethylsulphonyl chloride, Phenylmethanesulphonyl chloride

A132945-100G A132945-25G A132945-5G
ALA132945-100GEA 220.31 USD
ALA132945-100G ALA132945-25G ALA132945-5G
Toluene-α-sulfonyl chloride ≥97%
Toluene-α-sulphonyl chloride

Product Introduction:Phenylmethanesulfonyl chloride is an aliphatic sulfonyl chloride. It reacts with triethylamine in methylene chloride or ether to yield trans-stilbene and cis diphenylethylene sulfone. Its reaction with 1,1-bis(diethylamino)ethane has been studied. Addition of phenylmethanesulfonyl chloride to 1,3-diphenyl-2-pyrrolidinopropene in the presence of water was studied.

Formula: C₇H₇ClO₂S
MW: 190.65 g/mol
Boiling Pt: 120
Melting Pt: 92 °C
Storage Temperature: Ambient
MDL Number: MFCD00007455
CAS Number: 1939-99-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1b1723b587cd288205f3d1fe145089c5d5d569eb&vid=1b1723b587cd288205f3d1fe145089c5d5d569eb&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR