3'-Acetoxyacetophenone ≥98% (by GC)

Supplier: ALADDIN SCIENTIFIC
Warning

A132855-1G A132855-25G A132855-5G
ALA132855-1GEA 9.41 USD
ALA132855-1G ALA132855-25G ALA132855-5G
3'-Acetoxyacetophenone ≥98% (by GC)
3'-Acetoxyacetophenone
Formula: C₁₀H₁₀O₃
Boiling Pt: 147 °C
Melting Pt: 44.0 to 48.0 °C
MDL Number: MFCD00017227
CAS Number: 2454-35-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c6daf697c0d987baaadd8010f1bd08ea1d579bad&vid=c6daf697c0d987baaadd8010f1bd08ea1d579bad&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR