4-Aminobutyraldehyde dimethyl acetal ≥98%

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: 4-Aminobutyraldehyde Dimethyl Acetal, 4,4-Dimethoxybutylamine

A132534-1ML A132534-5ML
ALA132534-1MLEA 36.96 USD
ALA132534-1ML ALA132534-5ML
4-Aminobutyraldehyde dimethyl acetal ≥98%
4-Aminobutyraldehyde dimethyl acetal

ApplicProduct Application4-Aminobutyraldehyde dimethyl acetal acts as an intermediate in organic synthesis. It is used in the preparation of pineal hormone melatonin by reaction with 4-methoxyphenylhydrazine hydrochloride and acetic anhydride. Further, it is also used in the preparation of ficuseptine, juliprosine, and juliprosopine.

Formula: C₆H₁₅NO₂
Boiling Pt: 85 °C
Density: 0.95 g/cm³
Flash Pt: 59°C
Storage Temperature: Ambient
MDL Number: MFCD00189373
CAS Number: 19060-15-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=caf587b0d6de54615299c4c11ccd02f33a6000cb&vid=caf587b0d6de54615299c4c11ccd02f33a6000cb&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR