7-Aminoactinomycin D (7-AAD) ≥97% (by HPLC)

Supplier: ALADDIN SCIENTIFIC
Danger

A131295-1MG A131295-25MG A131295-5MG
ALA131295-1MGEA 112.01 USD
ALA131295-1MG ALA131295-25MG ALA131295-5MG
7-Aminoactinomycin D (7-AAD) ≥97% (by HPLC)
7-Aminoactinomycin D (7-AAD)

7-Aminoactinomycin D ,an analog of the antibiotic actinomycin D ,is a fluorescent DNA dye which is useful for quantifying apoptosis. 7-AAD has also been used in studies on cellular necrosis, chromosome analysis and in cell cycle studies. As a fluorescent DNA dye, 7-AAD is able to intercalate DNA via binding to the GC regions. When used in flow cytometery, 7-AAD can be used to discriminate between live cells as well as early and late apoptosis. Additionally, 7-AAD can also be combined with other staining procedures for analysis of intracellular and cell surface antigens enabling multiparametric analysis. When utilizing 7-AAD with flow cytometery, early and late apoptosis can be studied by varying the concentration of 7-AAD. During apoptosis, the cellular plasma membrane is altered progressively, which allows the cell membrane to become permeable to 7-AAD. 7-AAD staining can also be adapted to include staining with other dyes for intracellular and cell surface antigens. 7-Aminoactinomycin D is an inhibitor of DNA Polymerase and RNA PolymeraseA fluorescent DNA dye for apoptosis and flow cytomtery studies.

Formula: C₆₂H₈₇N₁₃O₁₆
Melting Pt: 252-253°C
Storage Temperature: Ambient
MDL Number: MFCD00077295
CAS Number: 7240-37-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6cd5c93ed9cd4c56e54a39fe2832a23d95bdb618&vid=6cd5c93ed9cd4c56e54a39fe2832a23d95bdb618&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR