4-Aminophthalic acid ≥95% (by titration)

Supplier: ALADDIN SCIENTIFIC
Warning

A130032-100G A130032-1G A130032-25G A130032-5G
ALA130032-100GEA 721.91 USD
ALA130032-100G ALA130032-1G ALA130032-25G ALA130032-5G
4-Aminophthalic acid ≥95% (by titration)
4-Aminophthalic acid
Formula: C₈H₇NO₄
Melting Pt: >300°C
Storage Temperature: Ambient
MDL Number: MFCD00013985
CAS Number: 5434-21-9
EINECS: 226-596-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7527b874a7f29a320528a9ea5288e97d36c9f7b4&vid=7527b874a7f29a320528a9ea5288e97d36c9f7b4&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR