Picolinamidine hydrochloride ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: Picolinimidamide hydrochloride, 2-Amidinopyridine hydrochloride

A130010-1G A130010-5G
ALA130010-1GEA 56.91 USD
ALA130010-1G ALA130010-5G
Picolinamidine hydrochloride ≥97%
Picolinamidine hydrochloride
Formula: C₆H₈ClN₃
MW: 157.60 g/mol
Melting Pt: 148-152°C
Storage Temperature: Ambient
MDL Number: MFCD00052271
CAS Number: 51285-26-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9f18055f3eb8abc40c77978e828c4ff68000bab0&vid=9f18055f3eb8abc40c77978e828c4ff68000bab0&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR