5-Methylpyrazine-2-carboxylic acid-4-oxide ≥98%, Moligand™

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: Acipimox, 5-Carboxy-2-methylpyrazine 1-oxide, 2-Carboxy-5-methylpyrazine 4-Oxide, 5-Carboxy-2-methylpyrazine 1-Oxide

A129989-1G A129989-25G A129989-5G
ALA129989-1GEA 74.01 USD
ALA129989-1G ALA129989-25G ALA129989-5G
5-Methylpyrazine-2-carboxylic acid-4-oxide ≥98%, Moligand™
5-Methylpyrazine-2-carboxylic acid-4-oxide

Acipimox is a niacin derivative.

Formula: C₆H₆N₂O₃
MW: 154.13 g/mol
Melting Pt: 180°C
Storage Temperature: Refrigerator
MDL Number: MFCD00865757
CAS Number: 51037-30-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1046585ffec3fbff59012556430ea8fd4771fbef&vid=1046585ffec3fbff59012556430ea8fd4771fbef&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR