2-Acetamido-5-nitrothiazole ≥98%
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
N-(5-Nitro-2-thiazolyl)acetamide, N-(5-Nitrothiazol-2-yl)acetamide
A129616-100G
A129616-1G
A129616-25G
A129616-5G
ALA129616-100GEA
2939.21
USD
ALA129616-100G
ALA129616-1G
ALA129616-25G
ALA129616-5G
2-Acetamido-5-nitrothiazole ≥98%
2-Acetamidot-5-nitrohiazole
Formula:
C₅H₅N₃O₃S MW: 187.18 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00022438 CAS Number: 140-40-9 EINECS: 205-414-7 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...