Azaperone ≥99%

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: 4'-Fluoro-4-[4-(2-pyridyl)-1-piperazinyl]butyrophenone, 1-(4-Fluorophenyl)-4-(4-(2-pyridinyl)-1-piperazinyl)-1-butanone

A129598-100MG A129598-1G A129598-250MG A129598-50MG
ALA129598100MGEA 84.46 USD
ALA129598100MG ALA129598-1G ALA129598250MG ALA129598-50MG
Azaperone ≥99%
Azaperone
Formula: C₁₉H₂₂FN₃₀
Melting Pt: 87-89°C
Storage Temperature: Refrigerator
MDL Number: MFCD00866692
CAS Number: 1649-18-9
EINECS: 216-715-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=679fa3cdc6a7bf2860d17b01a718efc6fb3d6974&vid=679fa3cdc6a7bf2860d17b01a718efc6fb3d6974&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR