Acemetacin ≥98%

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: [1-(4-Chlorobenzoyl)-5-methoxy-2-methylindol-3-yl]acetoxyacetic Acid, 2-(2-(1-(4-Chlorobenzoyl)-5-methoxy-2-methyl-1H-indol-3-yl)acetoxy)acetic acid, 1-(4-Chlorobenzoyl)-5-methoxy-2-methylindole-3-acetic Acid Carboxymethyl Ester

A129429-1G A129429-25G A129429-5G
ALA129429-1GEA 55.96 USD
ALA129429-1G ALA129429-25G ALA129429-5G
Acemetacin ≥98%
Acemetacin

Acemetacin is a glycolic acid ester of indometacin that is a cyclooxygenase inhibitor.

Formula: C₂₁H₁₈ClNO₆
Melting Pt: 152 °C
Storage Temperature: Freezer
MDL Number: MFCD00151473
CAS Number: 53164-05-9
EINECS: 258-403-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=592794d72a1855420b6d830bc96ebaba52b7fc6c&vid=592794d72a1855420b6d830bc96ebaba52b7fc6c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR