Alibendol ≥98%

Supplier: ALADDIN SCIENTIFIC

Synonyms: Cebera, 5-Allyl-2-hydroxy-N-(2-hydroxyethyl)-3-methoxybenzamide, 5-Allyl-N-(2-hydroxyethyl)-3-methoxysalicylamide

A129315-100G A129315-1G A129315-25G A129315-5G
ALA129315-100GEA 555.66 USD
ALA129315-100G ALA129315-1G ALA129315-25G ALA129315-5G
Alibendol ≥98%
Alibendol
Formula: C₁₃H₁₇NO₄
MW: 251.28 g/mol
Melting Pt: 100 °C
Storage Temperature: Freezer
MDL Number: MFCD00865165
CAS Number: 26750-81-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=92d75943f687c470ecaef1723ef8bbd2d18cedc9&vid=92d75943f687c470ecaef1723ef8bbd2d18cedc9&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR