Azilsartan ≥99%, Moligand™

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: 2-Ethoxy-1-((2'-(5-oxo-2,5-dihydro-1,2,4-oxadiazol-3-yl)-[1,1'-biphenyl]-4-yl)methyl)-1H-benzo[d]imidazole-7-carboxylic acid

A129255-10MG A129255-1G A129255-250MG A129255-25G A129255-50MG A129255-5G
ALA129255-10MGEA 61.66 USD
ALA129255-10MG ALA129255-1G ALA129255250MG ALA129255-25G ALA129255-50MG ALA129255-5G
Azilsartan ≥99%, Moligand™
Azilsartan
Formula: C₂₅H₂₀N₄O₅
Melting Pt: 188 °C
Storage Temperature: Refrigerator
MDL Number: MFCD20278186
CAS Number: 147403-03-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e128a02c4269aa3fcb75139131394b80e612c93d&vid=e128a02c4269aa3fcb75139131394b80e612c93d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR