AT-13387 ≥98%
Supplier: ALADDIN SCIENTIFIC
Synonyms:
(2,4-Dihydroxy-5-isopropylphenyl)(5-((4-methylpiperazin-1-yl)methyl)isoindolin-2-yl)methanone, Onalespib, AT13387, (2,4-dihydroxy-5-propan-2-ylphenyl)-[5-[(4-methylpiperazin-1-yl)methyl]-1,3-dihydroisoindol-2-yl]methanone, (2,4-dihydroxy-5-isopropylphenyl)(5-((4-methylpiperazin-1-yl)methyl)isoindolin-2-yl)methanone
A127900-10MG
A127900-5MG
ALA127900-10MGEA
94.91
USD
ALA127900-10MG
ALA127900-5MG
AT-13387 ≥98%
AT-13387
Formula:
C₂₄H₃₁N₃O₃ Storage Temperature: Freezer |
MDL Number:
MFCD18633198 CAS Number: 912999-49-6 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...