Apremilast ≥99%, Moligand™

Supplier: ALADDIN SCIENTIFIC
Danger

A126902-25G A126902-5G
ALA126902-25GEA 112817.16 USD
ALA126902-25G ALA126902-5G
Apremilast ≥99%, Moligand™
Apremilast

Apremilast (CC-10004) is a potent and orally active PDE4 and TNF-α inhibitor with IC50 of 74 nM and 77 nM, respectively.Apremilast(CC10004)是PDE4抑制剂,IC50为74 nM。

Formula: C₂₂H₂₄N₂O₇S
Storage Temperature: Freezer
MDL Number: MFCD18782607
CAS Number: 608141-41-9
EINECS: 807-237-6, 944-453-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1da7011f51bc239e7ed3c6f54627e514c24ad90b&vid=1da7011f51bc239e7ed3c6f54627e514c24ad90b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR