Ambrisentan 99, Moligand™

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: (S)-2-[(4,6-Dimethylpyrimidin-2-yl)oxy]-3-methoxy-3,3-diphenylpropionic acid, (S)-2-((4,6-Dimethylpyrimidin-2-yl)oxy)-3-methoxy-3,3-diphenylpropanoic acid

A126631-100MG A126631-10MG A126631-250MG A126631-25MG
ALA126631100MGEA 359.96 USD
ALA126631100MG ALA126631-10MG ALA126631250MG ALA126631-25MG
Ambrisentan 99, Moligand™
Ambrisentan

Ambrisentan is a selective ET type A receptor (ETAR) antagonist.

Formula: C₂₂H₂₂N₂O₄
MW: 378.42 g/mol
Melting Pt: 178 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09842330
CAS Number: 177036-94-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6db3c03461deb4ad325cf1f94507b391aacd5c25&vid=6db3c03461deb4ad325cf1f94507b391aacd5c25&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR