AZD3514 ≥98%

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: AZD 3514, 1-(4-(2-(4-(1-(7,8-dihydro-3-(trifluoromethyl)-1,2,4-triazolo(4,3-b)pyridazin-6-yl)-4-piperidinyl)phenoxy)ethyl)-1-piperazinyl)-ethanone, AZD-3514

A126496-100MG A126496-25MG A126496-5MG
ALA126496100MGEA 806.46 USD
ALA126496100MG ALA126496-25MG ALA126496-5MG
AZD3514 ≥98%
AZD3514

AZD3514 is a potent and oral androgen receptor downregulator with Ki of 2.2 μM and has ability of reducing AR protein expression.

Formula: C₂₅H₃₂F₃N₇O₂
Storage Temperature: Freezer
CAS Number: 1240299-33-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=13a72795285ef68a21d4f63c743909ae9446cba0&vid=13a72795285ef68a21d4f63c743909ae9446cba0&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR