AZD1480 ≥98%, Moligand™

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: (S)-5-Chloro-N2-(1-(5-fluoropyrimidin-2-yl)ethyl)-N4-(5-methyl-1H-pyrazol-3-yl)pyrimidine-2,4-diamine, AZD-1480

A126326-100MG A126326-10MG A126326-1MG A126326-25MG A126326-50MG A126326-5MG
ALA126326100MGEA 446.41 USD
ALA126326100MG ALA126326-10MG ALA126326-1MG ALA126326-25MG ALA126326-50MG ALA126326-5MG
AZD1480 ≥98%, Moligand™
AZD1480

AZD1480 has been used as an inhibitor of janus kinase 2 (JAK2) and signal transducer and activator of transcription 3 (STAT3) in an in vitro cell model of upper gastrointestinal adenocarcinoma. It has also been used in combination with bortezomib to study tumor-associated macrophage-mediated survival of myeloma cells.

Formula: C₁₄H₁₄ClFN₈
MW: 348.77 g/mol
Boiling Pt: ~575.2 °C
Melting Pt: 213.39°C
Storage Temperature: Freezer
MDL Number: MFCD16038904
CAS Number: 935666-88-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=34acb26ca701ca1644bded955c87315802304633&vid=34acb26ca701ca1644bded955c87315802304633&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR