Apixaban ≥99%, Moligand™

Supplier: ALADDIN SCIENTIFIC
Danger

A126046-10MG A126046-1G A126046-250MG A126046-50MG
ALA126046-10MGEA 68.31 USD
ALA126046-10MG ALA126046-1G ALA126046250MG ALA126046-50MG
Apixaban ≥99%, Moligand™
Apixaban

Apixaban is a highly selective, reversible inhibitor of Factor Xa with Ki of 0.08 nM and 0.17 nM in human and rabbit, respectively. Apixaban is in development for the prevention and treatment of various thromboembolic diseases.

Formula: C₂₅H₂₅N₅O₄
Storage Temperature: Freezer
MDL Number: MFCD11977295
CAS Number: 503612-47-3
EINECS: 639-684-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f09fc9191cedc5682d66988d650fff92c928a0b&vid=6f09fc9191cedc5682d66988d650fff92c928a0b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR