3-Amino-4-chloropyridine ≥98%

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: 4-Chloro-3-pyridylamine, 4-Chloropyridin-3-amine

A124742-5G A124742-100G A124742-1G A124742-25G
ALA124742-5GEA 103.46 USD
ALA124742-5G ALA124742-100G ALA124742-1G ALA124742-25G
3-Amino-4-chloropyridine ≥98%
3-Amino-4-chloropyridine
Formula: C₅H₅ClN₂
MW: 128.56 g/mol
Melting Pt: 60°C
Storage Temperature: Refrigerator
MDL Number: MFCD00235124
CAS Number: 20511-15-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=77aa95cabb9324146a256e42b2267f9b1facb77d&vid=77aa95cabb9324146a256e42b2267f9b1facb77d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR