Asenapine maleate ≥98% (by HPLC)

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: trans-5-Chloro-2,3,3a,12b-tetrahydro-2-methyl-1H-dibenz[2,3:6,7]oxepino[4,5-c]pyrrole maleate, Org 5222, (3aRS,12bRS)-5-Chloro-2-methyl-2,3,3a,12btetrahydro-1Hdibenzo[2,3:6,7]oxepino[4,5-c]pyrrole (2Z)-2-butenedioate

A124739-10MG A124739-1G A124739-250MG A124739-50MG
ALA124739-10MGEA 55.96 USD
ALA124739-10MG ALA124739-1G ALA124739250MG ALA124739-50MG
Asenapine maleate ≥98% (by HPLC)
Asenapine maleate

Asenapine maleate is a 5-HT (1A, 1B, 2A, 2B, 2C, 5A, 6, 7) and D2 antagonist with Ki values of 0.03-4.0 nM, 1.3nM, respectively, and an antipsychotic.Asenapine maleate is a serotonin 5-HT and dopamine D2 receptor antagonist. Asenapine maleate can be used to treat schizophrenia and bipolar mania.

Formula: C₁₇H₁₆ClNO ·C₄H₄O₄
Melting Pt: 141 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00900588
CAS Number: 85650-56-2
EINECS: 288-064-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=34a002dd42c3a30ad1c5c3c567fa873ace7335e8&vid=34a002dd42c3a30ad1c5c3c567fa873ace7335e8&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR