Agomelatine ≥98% (by HPLC), Moligand™
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
A124691-1G
A124691-250MG
A124691-25G
A124691-5G
ALA124691-1GEA
4633.06
USD
ALA124691-1G
ALA124691250MG
ALA124691-25G
ALA124691-5G
Agomelatine ≥98% (by HPLC), Moligand™
Agomelatine
Agomelatine is an extremely potent agonist at both melatonin receptors (MT1 and MT2), with additional antagonism at 5HT2C. It is a novel antidepressant with many desired in vivo properties, including neuroprotection and neurogenesis in depression-sensitive brain areas. Agomelatine′s efficacy appears to be due to both melatonergic and serotonergic properties. In neurogenesis assays, both in vitro and in vivo, the compound effects were differentially affected by antagonists for MT1/MT2 and 5HT2C, demonstrating actions through all three receptors.
Formula:
C₁₅H₁₇NO₂ Melting Pt: 110 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00916659 CAS Number: 138112-76-2 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...