4-Aminocyclohexanecarboxylic acid (mixture of cis and trans isomers) ≥95%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 4-Aminocyclohexanecarboxylic acid, 4-Aminocyclohexane-1-carboxylic acid

A124585-100G A124585-1G A124585-25G A124585-5G
ALA124585-100GEA 887.21 USD
ALA124585-100G ALA124585-1G ALA124585-25G ALA124585-5G
4-Aminocyclohexanecarboxylic acid (mixture of cis and trans isomers) ≥95%
4-Aminocyclohexanecarboxylic acid (mixture of cis and trans isomers)
Formula: C₇H₁₃NO₂
MW: 143.1800 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00191601
CAS Number: 1776-53-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1f9852c726d48b97cba062887e76a86fa8216352&vid=1f9852c726d48b97cba062887e76a86fa8216352&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR