2-Amino-3-nitro-5-fluoropyridine ≥98%

Supplier: ALADDIN SCIENTIFIC

A124341-1G A124341-250MG A124341-25G A124341-5G
ALA124341-1GEA 92.06 USD
ALA124341-1G ALA124341250MG ALA124341-25G ALA124341-5G
2-Amino-3-nitro-5-fluoropyridine ≥98%
2-Amino-3-nitro-5-fluoropyridine
Formula: C₅H₄FN₃O₂
MW: 157.1
MDL Number: MFCD04972413
CAS Number: 212268-12-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=100a1f747da59b1f154273e1bd9c67ef55b94e88&vid=100a1f747da59b1f154273e1bd9c67ef55b94e88&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR