3-Amino-5-chloropyridine ≥95%
Supplier: ALADDIN SCIENTIFIC
|
Danger
|
Synonyms:
5-Chloropyridin-3-amine
A124231-1G
A124231-250MG
A124231-5G
ALA124231-1GEA
42.66
USD
ALA124231-1G
ALA124231250MG
ALA124231-5G
3-Amino-5-chloropyridine ≥95%
3-Amino-5-chloropyridine
Formula:
C₅H₅ClN₂ MW: 128.56 Melting Pt: 71-75°C Storage Temperature: Ambient |
MDL Number:
MFCD03701386 CAS Number: 22353-34-0 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...