Amentoflavone ≥99% (by HPLC)

Supplier: ALADDIN SCIENTIFIC

Synonyms: I3′, Didemethyl-ginkgetin, Tridemethylsciadopitysin, II8-Biapigenin, 8-(5-(5,7-Dihydroxy-4-oxo-4H-chromen-2-yl)-2-hydroxyphenyl)-5,7-dihydroxy-2-(4-hydroxyphenyl)-4H-chromen-4-one

A123660-25MG A123660-5MG A123660-100MG
ALA123660-25MGEA 188.01 USD
ALA123660-25MG ALA123660-5MG ALA123660100MG
Amentoflavone ≥99% (by HPLC)
Amentoflavone

Biflavonoid with anti-inflammatory, anti-viral and cancer chemopreventive activity. It inhibits vascularization of tumors by blocking the activity of angiogenic VEGFs. Blocks the induction of COX-2 and up-regulates PPAR-γ. It is a negative modulator of the GABAA receptor at the benzodiazepine binding site.

Formula: C₃₀H₁₈O₁₀
MW: 538.464 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00017470
CAS Number: 1617-53-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f87d8b2ee9a839073791e4f8bc06d54159d645d0&vid=f87d8b2ee9a839073791e4f8bc06d54159d645d0&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR