Methyl-3-amino-2-methylbenzoate ≥96%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: Methyl 3-amino-2-methylbenzoate, 3-Amino-o-toluic acid Methyl Ester, 3-Amino-2-methylbenzoic acid Methyl Ester

A123493-100G A123493-1G A123493-25G A123493-5G
ALA123493-100GEA 143.36 USD
ALA123493-100G ALA123493-1G ALA123493-25G ALA123493-5G
Methyl-3-amino-2-methylbenzoate ≥96%
Methyl 3-amino-2-methylbenzoate
Formula: C₉H₁₁NO₂
Boiling Pt: 160°C
Density: 1.146 g/cm³
Flash Pt: 104°C
MDL Number: MFCD00211328
CAS Number: 18583-89-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11ecba3095cd3bdefecfa31289f068f21ac432d0&vid=11ecba3095cd3bdefecfa31289f068f21ac432d0&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR