5-Aminopyridine-2-carbonitrile ≥96%

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: 5-Amino-2-pyridinecarbonitrile, 5-Amino-2-cyanopyridine

A123440-100MG A123440-1G A123440-250MG
ALA123440100MGEA 9.41 USD
ALA123440100MG ALA123440-1G ALA123440250MG
5-Aminopyridine-2-carbonitrile ≥96%
5-Aminopyridine-2-carbonitrile
Formula: C₆H₅N₃
Melting Pt: 148-152°C
MDL Number: MFCD03093956
CAS Number: 55338-73-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7076bc4dbbfa2c29230a338cdccf9cb2c338bf3d&vid=7076bc4dbbfa2c29230a338cdccf9cb2c338bf3d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR