Agmatine sulfate ≥98%
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
1-Amino-4-guanidobutane Sulphate, 1-(4-Aminobutyl)guanidine sulfate, 1-(4-Aminobutyl)guanidine sulphate, 1-Amino-4-guanidobutane sulfate
A123125-100G
A123125-1G
A123125-25G
A123125-5G
ALA123125-100GEA
5471.91
USD
ALA123125-100G
ALA123125-1G
ALA123125-25G
ALA123125-5G
Agmatine sulfate ≥98%
Agmatine sulfate
Formula:
C₅H₁₄N₄ MW: 228.27 g/mol Melting Pt: 234-238°C |
MDL Number:
MFCD00013109 CAS Number: 2482-00-0 EINECS: 219-617-3 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...