2-Amino-6-fluorobenzonitrile ≥99%

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: 2-Cyano-3-fluoroaniline

A122708-100G A122708-1G A122708-25G A122708-5G
ALA122708-100GEA 695.31 USD
ALA122708-100G ALA122708-1G ALA122708-25G ALA122708-5G
2-Amino-6-fluorobenzonitrile ≥99%
2-Amino-6-fluorobenzonitrile

A medicinal chemistry intermediate used for the preparation of tacrine-related compounds and antifolate and antibacterial quinazoline derivatives.

Formula: C₇H₅FN₂
MW: 136.13
Boiling Pt: 272
Melting Pt: 127
Storage Temperature: Refrigerator
MDL Number: MFCD00015477
CAS Number: 77326-36-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32879e70ac923767b3a88746ab9fc4f2d429d148&vid=32879e70ac923767b3a88746ab9fc4f2d429d148&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR