4-Amino-2-chlorobenzonitrile ≥99%

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: 3-Chloro-4-cyanoaniline

A122704-100G A122704-25G A122704-5G
ALA122704-100GEA 229.81 USD
ALA122704-100G ALA122704-25G ALA122704-5G
4-Amino-2-chlorobenzonitrile ≥99%
4-Amino-2-chlorobenzonitrile
Formula: C₇H₅ClN₂
MW: 152.58 g/mol
Boiling Pt: 301
Melting Pt: 116-119°C
MDL Number: MFCD00035926
CAS Number: 20925-27-3
EINECS: 244-114-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=efb8e6d627ceae8f2330d56f632825b167362add&vid=efb8e6d627ceae8f2330d56f632825b167362add&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR