2-Amino-4,5-dimethylthiazole hydrochloride 98

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: 4,5-Dimethylthiazol-2-amine hydrochloride

A122571-25G A122571-5G
ALA122571-25GEA 565.16 USD
ALA122571-25G ALA122571-5G
2-Amino-4,5-dimethylthiazole hydrochloride 98
2-Amino-4,5-dimethylthiazole hydrochloride

Used in the synthesis of 2-[3-[3-(piperidinomethyl?)phenoxy]propionamido]-4,?5-dimethylthiazole.

Formula: C₅H₉ClN₂S
MW: 164.66 g/mol
Melting Pt: 273-274°C
MDL Number: MFCD00012711
CAS Number: 71574-33-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6551e6d898db876df177b079f401358608129031&vid=6551e6d898db876df177b079f401358608129031&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR